Product Image
Thermo Scientific™

UltiMate™ MWD-3000RS to DAD-3000RS Upgrade Kit

Capture more information from your sample by using this kit to upgrade your Thermo Scientific™ Dionex™ UltiMate™ MWD-3000RS detector to a DAD 3000RS detector.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
6082.3035Upgrade Kit (MWD-3000 to DAD-3000)Dionex Ultimate MWD-3000
6082.3045Upgrade Kit (MWD-3000RS to DAD-3000RS)Dionex Ultimate MWD-3000 RS
Catalog number 6082.3035
Price (USD)
-
Type:
Upgrade Kit (MWD-3000 to DAD-3000)
For Use With (Equipment):
Dionex Ultimate MWD-3000
Capture more information from samples analyzed by LC. Use the Thermo Scientific™ Dionex™ UltiMate™ MWD-3000RS to DAD-3000RS Upgrade kit to upgrade from the multiwavelength detector to the diode array detector. This kit includes all components for upgrading an UltiMate MWD-3000 to an UltiMate DAD-3000 detector. The kit must be installed by authorized service personnel.
Specifications
Detection ModeUV-vis Detection (8 Channels)
Drift<0.5 mAU/hr
For Use With (Equipment)Dionex Ultimate MWD-3000
GLP FeaturesAutomatic Instrument Qualification (AutoQ), System Wellness, and Qualification Monitoring with Chromeleon software, lamp and cell ID chips
Linearity<3%RSD
Noise< ±7 μAU
PC ConnectionUSB 2.0
Pixel Resolution<1 nm
Slit WidthNarrow or Wide
Voltage85 to 260 V
Wavelength Accuracy±1.0 nm
Wavelength Range190 to 800 nm
Data Collection Rate100 Hz Max.
Detector TypeSingle-Beam
TypeUpgrade Kit (MWD-3000 to DAD-3000)
Unit SizeEach