CD3e Monoclonal Antibody (145-2C11), PE-eFluor™ 610, eBioscience™ - FAQs
CD3e Monoclonal Antibody (145-2C11), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD3e Monoclonal Antibody (145-2C11), PE-eFluor™ 610, eBioscience™ - FAQs (61-0031-80, 61-0031-82)