CD3 Monoclonal Antibody (UCHT1), PE-eFluor™ 610, eBioscience™ - FAQs
CD3 Monoclonal Antibody (UCHT1), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD3 Monoclonal Antibody (UCHT1), PE-eFluor™ 610, eBioscience™ - FAQs (61-0038-41, 61-0038-42)