CD4 Monoclonal Antibody (GK1.5), PE-eFluor™ 610, eBioscience™ - FAQs
CD4 Monoclonal Antibody (GK1.5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (GK1.5), PE-eFluor™ 610, eBioscience™ - FAQs (61-0041-80, 61-0041-82)