CD4 Monoclonal Antibody (RM4-5), PE-eFluor™ 610, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RM4-5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RM4-5), PE-eFluor™ 610, eBioscience™ - FAQs (61-0042-82, 61-0042-80)