CD4 Monoclonal Antibody (RPA-T4), PE-eFluor™ 610, eBioscience™ - FAQs
CD4 Monoclonal Antibody (RPA-T4), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD4 Monoclonal Antibody (RPA-T4), PE-eFluor™ 610, eBioscience™ - FAQs (61-0049-42, 61-0049-41)