CD8a Monoclonal Antibody (53-6.7), PE-eFluor™ 610, eBioscience™ - FAQs
CD8a Monoclonal Antibody (53-6.7), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (53-6.7), PE-eFluor™ 610, eBioscience™ - FAQs (61-0081-80, 61-0081-82)