CD8a Monoclonal Antibody (RPA-T8), PE-eFluor™ 610, eBioscience™ - FAQs
CD8a Monoclonal Antibody (RPA-T8), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD8a Monoclonal Antibody (RPA-T8), PE-eFluor™ 610, eBioscience™ - FAQs (61-0088-42, 61-0088-41)