CD11b Monoclonal Antibody (M1/70), PE-eFluor™ 610, eBioscience™ - FAQs
CD11b Monoclonal Antibody (M1/70), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD11b Monoclonal Antibody (M1/70), PE-eFluor™ 610, eBioscience™ - FAQs (61-0112-82, 61-0112-80)