CD11c Monoclonal Antibody (N418), PE-eFluor™ 610, eBioscience™ - FAQs
CD11c Monoclonal Antibody (N418), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (N418), PE-eFluor™ 610, eBioscience™ - FAQs (61-0114-80, 61-0114-82)