CD11c Monoclonal Antibody (3.9), PE-eFluor™ 610, eBioscience™ - FAQs
CD11c Monoclonal Antibody (3.9), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD11c Monoclonal Antibody (3.9), PE-eFluor™ 610, eBioscience™ - FAQs (61-0116-42)