CD14 Monoclonal Antibody (61D3), PE-eFluor™ 610, eBioscience™ - FAQs
CD14 Monoclonal Antibody (61D3), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD14 Monoclonal Antibody (61D3), PE-eFluor™ 610, eBioscience™ - FAQs (61-0149-42, 61-0149-41)