CD19 Monoclonal Antibody (eBio1D3 (1D3)), PE-eFluor™ 610, eBioscience™ - FAQs
CD19 Monoclonal Antibody (eBio1D3 (1D3)), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (eBio1D3 (1D3)), PE-eFluor™ 610, eBioscience™ - FAQs (61-0193-82, 61-0193-80)