CD19 Monoclonal Antibody (HIB19), PE-eFluor™ 610, eBioscience™ - FAQs
CD19 Monoclonal Antibody (HIB19), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD19 Monoclonal Antibody (HIB19), PE-eFluor™ 610, eBioscience™ - FAQs (61-0199-41, 61-0199-42)