CD20 Monoclonal Antibody (2H7), PE-eFluor™ 610, eBioscience™ - FAQs
CD20 Monoclonal Antibody (2H7), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD20 Monoclonal Antibody (2H7), PE-eFluor™ 610, eBioscience™ - FAQs (61-0209-41, 61-0209-42)