CD25 Monoclonal Antibody (PC61.5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (PC61.5), PE-eFluor™ 610, eBioscience™ - FAQs (61-0251-82, 61-0251-80)