CD25 Monoclonal Antibody (CD25-4E3), PE-eFluor™ 610, eBioscience™ - FAQs
CD25 Monoclonal Antibody (CD25-4E3), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD25 Monoclonal Antibody (CD25-4E3), PE-eFluor™ 610, eBioscience™ - FAQs (61-0257-42)