CD27 Monoclonal Antibody (O323), PE-eFluor™ 610, eBioscience™ - FAQs
CD27 Monoclonal Antibody (O323), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD27 Monoclonal Antibody (O323), PE-eFluor™ 610, eBioscience™ - FAQs (61-0279-42, 61-0279-41)