CD28 Monoclonal Antibody (CD28.2), PE-eFluor™ 610, eBioscience™ - FAQs
CD28 Monoclonal Antibody (CD28.2), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD28 Monoclonal Antibody (CD28.2), PE-eFluor™ 610, eBioscience™ - FAQs (61-0289-41, 61-0289-42)