CD34 Monoclonal Antibody (4H11), PE-eFluor™ 610, eBioscience™ - FAQs
CD34 Monoclonal Antibody (4H11), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD34 Monoclonal Antibody (4H11), PE-eFluor™ 610, eBioscience™ - FAQs (61-0349-42, 61-0349-41)