CD38 Monoclonal Antibody (HIT2), PE-eFluor™ 610, eBioscience™ - FAQs
CD38 Monoclonal Antibody (HIT2), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD38 Monoclonal Antibody (HIT2), PE-eFluor™ 610, eBioscience™ - FAQs (61-0389-42, 61-0389-41)