CD44 Monoclonal Antibody (IM7), PE-eFluor™ 610, eBioscience™ - FAQs
CD44 Monoclonal Antibody (IM7), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD44 Monoclonal Antibody (IM7), PE-eFluor™ 610, eBioscience™ - FAQs (61-0441-82, 61-0441-80)