CD45 Monoclonal Antibody (30-F11), PE-eFluor™ 610, eBioscience™ - FAQs
CD45 Monoclonal Antibody (30-F11), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (30-F11), PE-eFluor™ 610, eBioscience™ - FAQs (61-0451-82, 61-0451-80)