CD45.1 Monoclonal Antibody (A20), PE-eFluor™ 610, eBioscience™ - FAQs
CD45.1 Monoclonal Antibody (A20), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD45.1 Monoclonal Antibody (A20), PE-eFluor™ 610, eBioscience™ - FAQs (61-0453-80, 61-0453-82)