CD45.2 Monoclonal Antibody (104), PE-eFluor™ 610, eBioscience™ - FAQs
CD45.2 Monoclonal Antibody (104), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD45.2 Monoclonal Antibody (104), PE-eFluor™ 610, eBioscience™ - FAQs (61-0454-82, 61-0454-80)