CD45RA Monoclonal Antibody (HI100), PE-eFluor™ 610, eBioscience™ - FAQs
CD45RA Monoclonal Antibody (HI100), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD45RA Monoclonal Antibody (HI100), PE-eFluor™ 610, eBioscience™ - FAQs (61-0458-41, 61-0458-42)