CD45 Monoclonal Antibody (HI30), PE-eFluor™ 610, eBioscience™ - FAQs
CD45 Monoclonal Antibody (HI30), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD45 Monoclonal Antibody (HI30), PE-eFluor™ 610, eBioscience™ - FAQs (61-0459-41, 61-0459-42)