CD117 (c-Kit) Monoclonal Antibody (2B8), PE-eFluor™ 610, eBioscience™ - FAQs
CD117 (c-Kit) Monoclonal Antibody (2B8), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD117 (c-Kit) Monoclonal Antibody (2B8), PE-eFluor™ 610, eBioscience™ - FAQs (61-1171-82)