CD123 Monoclonal Antibody (6H6), PE-eFluor™ 610, eBioscience™ - FAQs
CD123 Monoclonal Antibody (6H6), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD123 Monoclonal Antibody (6H6), PE-eFluor™ 610, eBioscience™ - FAQs (61-1239-42)