CD127 Monoclonal Antibody (eBioRDR5), PE-eFluor™ 610, eBioscience™ - FAQs
CD127 Monoclonal Antibody (eBioRDR5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD127 Monoclonal Antibody (eBioRDR5), PE-eFluor™ 610, eBioscience™ - FAQs (61-1278-41, 61-1278-42)