CD133 (Prominin-1) Monoclonal Antibody (TMP4), PE-eFluor™ 610, eBioscience™ - FAQs
CD133 (Prominin-1) Monoclonal Antibody (TMP4), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD133 (Prominin-1) Monoclonal Antibody (TMP4), PE-eFluor™ 610, eBioscience™ - FAQs (61-1338-41, 61-1338-42)