CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PE-eFluor™ 610, eBioscience™ - FAQs
CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD154 (CD40 Ligand) Monoclonal Antibody (24-31), PE-eFluor™ 610, eBioscience™ - FAQs (61-1548-42, 61-1548-41)