CD161 Monoclonal Antibody (HP-3G10), PE-eFluor™ 610, eBioscience™ - FAQs
CD161 Monoclonal Antibody (HP-3G10), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD161 Monoclonal Antibody (HP-3G10), PE-eFluor™ 610, eBioscience™ - FAQs (61-1619-41, 61-1619-42)