CD169 (Siglec-1) Monoclonal Antibody (7-239), PE-eFluor™ 610, eBioscience™ - FAQs
CD169 (Siglec-1) Monoclonal Antibody (7-239), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD169 (Siglec-1) Monoclonal Antibody (7-239), PE-eFluor™ 610, eBioscience™ - FAQs (61-1699-41, 61-1699-42)