CD196 (CCR6) Monoclonal Antibody (R6H1), PE-eFluor™ 610, eBioscience™ - FAQs
CD196 (CCR6) Monoclonal Antibody (R6H1), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD196 (CCR6) Monoclonal Antibody (R6H1), PE-eFluor™ 610, eBioscience™ - FAQs (61-1969-42)