CD197 (CCR7) Monoclonal Antibody (3D12), PE-eFluor™ 610, eBioscience™ - FAQs
CD197 (CCR7) Monoclonal Antibody (3D12), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD197 (CCR7) Monoclonal Antibody (3D12), PE-eFluor™ 610, eBioscience™ - FAQs (61-1979-42, 61-1979-41)