CD205 Monoclonal Antibody (205yekta), PE-eFluor™ 610, eBioscience™ - FAQs
CD205 Monoclonal Antibody (205yekta), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD205 Monoclonal Antibody (205yekta), PE-eFluor™ 610, eBioscience™ - FAQs (61-2051-82, 61-2051-80)