CD223 (LAG-3) Monoclonal Antibody (3DS223H), PE-eFluor™ 610, eBioscience™ - FAQs
CD223 (LAG-3) Monoclonal Antibody (3DS223H), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD223 (LAG-3) Monoclonal Antibody (3DS223H), PE-eFluor™ 610, eBioscience™ - FAQs (61-2239-42, 61-2239-41)