CD279 (PD-1) Monoclonal Antibody (eBioJ105 (J105)), PE-eFluor™ 610, eBioscience™ - FAQs
CD279 (PD-1) Monoclonal Antibody (eBioJ105 (J105)), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD279 (PD-1) Monoclonal Antibody (eBioJ105 (J105)), PE-eFluor™ 610, eBioscience™ - FAQs (61-2799-41, 61-2799-42)