CD317 (BST2, PDCA-1) Monoclonal Antibody (eBio927), PE-eFluor™ 610, eBioscience™ - FAQs
CD317 (BST2, PDCA-1) Monoclonal Antibody (eBio927), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD317 (BST2, PDCA-1) Monoclonal Antibody (eBio927), PE-eFluor™ 610, eBioscience™ - FAQs (61-3172-82, 61-3172-80)