CD335 (NKp46) Monoclonal Antibody (29A1.4), PE-eFluor™ 610, eBioscience™ - FAQs
CD335 (NKp46) Monoclonal Antibody (29A1.4), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for CD335 (NKp46) Monoclonal Antibody (29A1.4), PE-eFluor™ 610, eBioscience™ - FAQs (61-3351-80, 61-3351-82)