Rat anti-Mouse IgG1 Secondary Antibody, PE-eFluor™ 610, eBioscience™ - FAQs
Rat anti-Mouse IgG1 Secondary Antibody, PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Rat anti-Mouse IgG1 Secondary Antibody, PE-eFluor™ 610, eBioscience™ - FAQs (61-4015-80, 61-4015-82)