Rat IgG2b kappa Isotype Control (eB149/10H5), PE-eFluor™ 610, eBioscience™ - FAQs
Rat IgG2b kappa Isotype Control (eB149/10H5), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Rat IgG2b kappa Isotype Control (eB149/10H5), PE-eFluor™ 610, eBioscience™ - FAQs (61-4031-80)