Rat IgG1 kappa Isotype Control (eBRG1), PE-eFluor™ 610, eBioscience™ - FAQs
Rat IgG1 kappa Isotype Control (eBRG1), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Rat IgG1 kappa Isotype Control (eBRG1), PE-eFluor™ 610, eBioscience™ - FAQs (61-4301-80, 61-4301-82)