eBioscience™ Streptavidin PE-eFluor™ 610 Conjugate - FAQs
eBioscience™ Streptavidin PE-eFluor™ 610 Conjugate - FAQs
View additional product information for eBioscience™ Streptavidin PE-eFluor™ 610 Conjugate - FAQs (61-4317-82, 61-4317-80)