Rat IgG2a kappa Isotype Control (eBR2a), PE-eFluor™ 610, eBioscience™ - FAQs
Rat IgG2a kappa Isotype Control (eBR2a), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Rat IgG2a kappa Isotype Control (eBR2a), PE-eFluor™ 610, eBioscience™ - FAQs (61-4321-82, 61-4321-80)