Rat IgM Isotype Control (eBRM), PE-eFluor™ 610, eBioscience™ - FAQs
Rat IgM Isotype Control (eBRM), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Rat IgM Isotype Control (eBRM), PE-eFluor™ 610, eBioscience™ - FAQs (61-4341-80)