Mouse IgG2a kappa Isotype Control (eBM2a), PE-eFluor™ 610, eBioscience™ - FAQs
Mouse IgG2a kappa Isotype Control (eBM2a), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Mouse IgG2a kappa Isotype Control (eBM2a), PE-eFluor™ 610, eBioscience™ - FAQs (61-4724-80)