Mouse IgG2b kappa Isotype Control (eBMG2b), PE-eFluor™ 610, eBioscience™ - FAQs
Mouse IgG2b kappa Isotype Control (eBMG2b), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for Mouse IgG2b kappa Isotype Control (eBMG2b), PE-eFluor™ 610, eBioscience™ - FAQs (61-4732-80)