FOXP3 Monoclonal Antibody (PCH101), PE-eFluor™ 610, eBioscience™ - FAQs
FOXP3 Monoclonal Antibody (PCH101), PE-eFluor™ 610, eBioscience™ - FAQs
View additional product information for FOXP3 Monoclonal Antibody (PCH101), PE-eFluor™ 610, eBioscience™ - FAQs (61-4776-41, 61-4776-42)